accessibility menu, dialog, popup

UserName

CH3H3CNO

CAS RN 1073-23-0

IUPAC Name: 2,6-dimethylpyridin-1-ium-1-olate
Synonyms: 2,6-dimethylpyridin-1-ium-1-olate
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: LIDGFHXPUOJZMK-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=[N+]1[O-]
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)