missing translation for 'onlineSavingsMsg'
Learn More

2,6-Lutidine N-oxide, 98%

Catalog No. AAA1429930
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1429930 250 g
AAA1429909 10 g
AAA1429918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1429930 Supplier Thermo Scientific Chemicals Supplier No. A1429930
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1073-23-0 | C7H9NO | 123.155 g/mol

2,6-Lutidine N-oxide is used as a ligand and form coordination complexes with metal salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Lutidine N-oxide is used as a ligand and form coordination complexes with metal salts.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with oxidizing agents, moisture and water.

Chemical Identifiers

CAS 1073-23-0
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00039715
InChI Key LIDGFHXPUOJZMK-UHFFFAOYSA-N
Synonym 2,6-dimethylpyridine n-oxide, 2,6-lutidine n-oxide, 2,6-lutidine-n-oxide, 2,6-dimethylpyridine 1-oxide, 2,6-lutidine 1-oxide, pyridine, 2,6-dimethyl-, 1-oxide, 2,6-lutidine oxide, 2,6-dimethylpyridine oxide, 2,6-dimethylpyridinium n-oxide, 2,6-dimethylpyridin-1-ium-1-olate
PubChem CID 14083
IUPAC Name 2,6-dimethyl-1-oxidopyridin-1-ium
SMILES CC1=[N+](C(=CC=C1)C)[O-]

Specifications

Density 1.102
Boiling Point 108°C to 111°C (10 mmHg)
Refractive Index 1.5685
Quantity 250 g
Beilstein 110551
Sensitivity Hygroscopic
Solubility Information Miscible with water.
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material 2,6-Lutidine N-oxide
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 214-025-1
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only