accessibility menu, dialog, popup

UserName

CAS RN 1072-98-6

IUPAC Name: 5-chloropyridin-2-amine
Synonyms: 2-amino-5-chloropyridine
Molecular Weight (g/mol): 128.56
Molecular Formula: C5H5ClN2
InChi Key: MAXBVGJEFDMHNV-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C=N1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (3)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)