missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloropyridine, 98%

Catalog No. AC103415000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103415000 500 g Plastic bottle
AC103410250 25 g Plastic bottle
AC103411000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC103415000 Supplier Thermo Scientific Chemicals Supplier No. 103415000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1072-98-6 | C5H5ClN2 | 128.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1072-98-6
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.56
MDL Number MFCD00006324
InChI Key MAXBVGJEFDMHNV-UHFFFAOYSA-N
Synonym 2-amino-5-chloropyridine, 2-pyridinamine, 5-chloro, 5-chloro-2-pyridinamine, 5-chloro-2-pyridylamine, 5-chloro-2-aminopyridine, 5-chloro-pyridin-2-ylamine, amino 2--5-chloropyridine, pyridine, 2-amino-5-chloro, 2-amino-5-chloro pyridine, 2-amino5-chloropyridine
PubChem CID 66174
IUPAC Name 5-chloropyridin-2-amine
SMILES C1=CC(=NC=C1Cl)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 134.0°C to 138.0°C
Color Beige to Beige-Brown
Boiling Point 127.0°C to 128.0°C (11.0 mmHg)
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula Cl(C5H3N)NH2
Quantity 500 g
Beilstein 22, V,8, 451
Solubility Information Solubility in water: 1g/L water (20°C)
Formula Weight 128.56
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-chloropyridine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
EINECSNumber 214-020-4

RUO – Research Use Only