missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-chloropyridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 136°C |
| Quantity | 500 g |
| Formula Weight | 128.56 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Amino-5-chloropyridine |
Safety and Handling
| TSCA | No |