accessibility menu, dialog, popup

UserName

H2NOHO

CAS RN 107-95-9

IUPAC Name: 3-aminopropanoic acid
Synonyms: β alanine|bAla|ω-aminopropionate|abufene|H-β-ala-OH|β-aminopropionsaeure|3-aminopropionsaeure|Β-alanine|ω-aminopropionic acid|monosodium salt β alanine
Molecular Weight (g/mol): 89.09
Molecular Formula: C3H7NO2
InChi Key: UCMIRNVEIXFBKS-UHFFFAOYSA-N
SMILES: NCCC(O)=O
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)