accessibility menu, dialog, popup

UserName

CAS RN 1069-66-5

NaH3CH3COO

CAS RN 1069-66-5

IUPAC Name: sodium 2-propylpentanoate
Synonyms: sodium, valproate epilim depakene
Molecular Weight (g/mol): 166.2
Molecular Formula: C8H15NaO2
InChi Key: AEQFSUDEHCCHBT-UHFFFAOYSA-M
SMILES: [Na+].CCCC(CCC)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results