accessibility menu, dialog, popup

UserName

CH3H3CCH3OONH(R)H2NOOH

CAS RN 106719-44-2

IUPAC Name: (2R)-6-amino-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid
Synonyms: (2R)-6-amino-2-[(tert-butoxycarbonyl)amino]hexanoic acid
Molecular Weight (g/mol): 246.31
Molecular Formula: C11H22N2O4
InChi Key: DQUHYEDEGRNAFO-SVGMAFHSNA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CCCCN)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)