missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-D-lysine, 98%

Catalog No. AAH6296606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6296606 5 g
AAH6296603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6296606 Supplier Thermo Scientific Chemicals Supplier No. H6296606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 106719-44-2 | C11H22N2O4 | 246.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106719-44-2
Molecular Formula C11H22N2O4
Molecular Weight (g/mol) 246.31
MDL Number MFCD00076956
InChI Key DQUHYEDEGRNAFO-SVGMAFHSNA-N
Synonym boc-d-lys-oh, boc-l-lys-oh, n2-boc-d-lysine, nalpha-tert-butoxycarbonyl-d-lysine, nalpha-boc-d-lysine, n-, a-boc-d-lysine, boc-d-lysine, d-lysine, n2-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl-d-lysine, n-alpha-t-butoxycarbonyl-d-lysine
PubChem CID 7018770
IUPAC Name (2R)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CCCCN)C(O)=O

Specifications

Melting Point 196°C to 200°C
Quantity 5 g
Optical Rotation −22° (c=2 in Methanol)
Formula Weight 246.3
Percent Purity 98%
Chemical Name or Material Nalpha-Boc-D-lysine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only