missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-(tert-Butoxycarbonyl)-D-lysine 98.0+%, TCI America™

Catalog No. B30835G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30835G 5 g
B30831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30835G Supplier TCI America Supplier No. B30835G

Chemical Identifiers

CAS 106719-44-2
Molecular Formula C11H22N2O4
Molecular Weight (g/mol) 246.31
MDL Number MFCD00076956
InChI Key DQUHYEDEGRNAFO-SVGMAFHSNA-N
Synonym boc-d-lys-oh, boc-l-lys-oh, n2-boc-d-lysine, nalpha-tert-butoxycarbonyl-d-lysine, nalpha-boc-d-lysine, n-, a-boc-d-lysine, boc-d-lysine, d-lysine, n2-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl-d-lysine, n-alpha-t-butoxycarbonyl-d-lysine
PubChem CID 7018770
IUPAC Name (2R)-6-amino-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CCCCN)C(O)=O

Specifications

Melting Point 198°C
Color White
Quantity 5 g
Formula Weight 246.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(tert-Butoxycarbonyl)-D-lysine