accessibility menu, dialog, popup

UserName

CAS RN 106483-04-9

H3COONOOCH3OHCH3

CAS RN 106483-04-9

IUPAC Name: {3-hydroxy-2-methyl-5-[(octanoyloxy)methyl]pyridin-4-yl}methyl octanoate
Synonyms: pyridoxine dicaprylate
Molecular Weight (g/mol): 421.58
Molecular Formula: C24H39NO5
InChi Key: PAUSGZCRNOTKPK-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)OCC1=CN=C(C)C(O)=C1COC(=O)CCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results