accessibility menu, dialog, popup

UserName

H3CONN

CAS RN 106144-18-7

IUPAC Name: 4-propoxybenzene-1,2-dicarbonitrile
Synonyms: 4-propoxybenzene-1,2-dicarbonitrile
Molecular Weight (g/mol): 186.21
Molecular Formula: C11H10N2O
InChi Key: RDLZRCSOLOTEPK-UHFFFAOYSA-N
SMILES: CCCOC1=CC(C#N)=C(C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results