accessibility menu, dialog, popup

UserName

NH2SNH2N

CAS RN 106092-09-5

IUPAC Name: 4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine
Synonyms: 4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine
Molecular Weight (g/mol): 169.25
Molecular Formula: C7H11N3S
InChi Key: DRRYZHHKWSHHFT-UHFFFAOYNA-N
SMILES: NC1CCC2=C(C1)SC(N)=N2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results