accessibility menu, dialog, popup

UserName

CAS RN 106-58-1

IUPAC Name: 1,4-dimethylpiperazine
Synonyms: piperazine, 1,4-dimethyl-
Molecular Weight (g/mol): 114.19
Molecular Formula: C6H14N2
InChi Key: RXYPXQSKLGGKOL-UHFFFAOYSA-N
SMILES: CN1CCN(C)CC1
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)