missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethylpiperazine, 98%

Catalog No. AAB2441922
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2441922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2441922 Supplier Thermo Scientific Chemicals Supplier No. B2441922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 106-58-1 | C6H14N2 | 114.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-58-1
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00006156
InChI Key RXYPXQSKLGGKOL-UHFFFAOYSA-N
Synonym n,n'-dimethylpiperazine, lupetazine, piperazine, 1,4-dimethyl, texacat dmp, unii-tf146u602l, ccris 6690, 1,4-dimethyl-piperazine, 1,4-dimethyl-2,3,5,6-tetrahydropyrazine, 1,4-dimethylpiperazin, pubchem8569
PubChem CID 7818
IUPAC Name 1,4-dimethylpiperazine
SMILES CN1CCN(CC1)C

Specifications

Melting Point -1°C
Density 0.844
pH 10
Boiling Point 131°C to 132°C
Flash Point 21°C (69°F)
Odor Amine-like
Refractive Index 1.447
Quantity 100 g
UN Number UN2733
Beilstein 103022
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material 1,4-Dimethylpiperazine
Show More Show Less
Hazard Category H225-H290-H302-H314-H318
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P234-P240-P241-P242-P243-P260-P264b-P270-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 203-412-0
RTECSNumber TL5945000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only