missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethylpiperazine 98.0+%, TCI America™

Catalog No. D0787500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0787500ML 500 mL
D078725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0787500ML Supplier TCI America Supplier No. D0787500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106-58-1
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00006156
InChI Key RXYPXQSKLGGKOL-UHFFFAOYSA-N
Synonym n,n'-dimethylpiperazine, lupetazine, piperazine, 1,4-dimethyl, texacat dmp, unii-tf146u602l, ccris 6690, 1,4-dimethyl-piperazine, 1,4-dimethyl-2,3,5,6-tetrahydropyrazine, 1,4-dimethylpiperazin, pubchem8569
PubChem CID 7818
IUPAC Name 1,4-dimethylpiperazine
SMILES CN1CCN(CC1)C

Specifications

Melting Point -1°C
Color Yellow
Boiling Point 133°C
Quantity 500 mL
UN Number 2924
Formula Weight 114.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N′-Dimethylpiperazine
EINECSNumber (5)-7024
RTECSNumber TL5945000
TSCA Yes