accessibility menu, dialog, popup

UserName

CAS RN 106-33-2

IUPAC Name: ethyl dodecanoate
Synonyms: ethyl laurate
Molecular Weight (g/mol): 228.38
Molecular Formula: C14H28O2
InChi Key: MMXKVMNBHPAILY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(=O)OCC
Percent Purity 
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)