missing translation for 'onlineSavingsMsg'
Learn More

Ethyl laurate, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012394
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012394 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012394 Supplier Merck Emd Millipore Supplier No. 075231ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 106-33-2
Density 0.862 g/mL (at 20°C)
Flash Point 113°C
Linear Formula CH3(CH2)10 COOC2H5
MDL Number MFCD00015065
Boiling Point 269°C
Synonym Ethyl dodecanoate; Ethyl laurate; Lauric acid ethyl ester
Type Analytical Standard
Quantity 1 mL
Formula Weight 228.37
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less