CAS RN 10534-89-1
IUPAC Name:
cobalt(3+) hexaamine trichloride
Synonyms:
hexamminecobalt(III) chloride
Molecular Weight (g/mol):
267.47
Molecular Formula:
Cl3CoH18N6
InChi Key:
JXBGZYGSWFSYFI-UHFFFAOYSA-K
SMILES:
N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Co+3]
1
–
5
of
5
results
1
Hexaamminecobalt(III) chloride, 97%
CAS: 10534-89-1 Molecular Formula: [Co(NH3)6]Cl3 MDL Number: MFCD00036304
2
Hexaamminecobalt(III) chloride, 99%
CAS: 10534-89-1 Molecular Formula: H18Cl3CoN6 MDL Number: MFCD00036304
3
Hexaamminecobalt(III) chloride, 99%
CAS: 10534-89-1 Molecular Formula: H18Cl3CoN6 Molecular Weight (g/mol): 267.47 MDL Number: MFCD00036304
4
Hexaamminecobalt(III) chloride, 99.999%, (trace metal basis)
CAS: 10534-89-1 Molecular Formula: H18Cl3CoN6 Molecular Weight (g/mol): 267.47 MDL Number: MFCD00036304
5
Hexaamminecobalt(III) Chloride 99.0+%, TCI America™
CAS: 10534-89-1 Molecular Formula: H18Cl3CoN6 MDL Number: MFCD00036304 Synonym: Luteo-cobaltic Chloride