missing translation for 'onlineSavingsMsg'
Learn More

Hexaamminecobalt(III) chloride, 99%

Catalog No. AC405311000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC405311000 100 g Glass bottle
AC405310050 5 g Glass bottle
AC405310250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC405311000 Supplier Thermo Scientific Chemicals Supplier No. 405311000

CAS: 10534-89-1 | H18Cl3CoN6 | 267.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10534-89-1
Molecular Formula H18Cl3CoN6
Molecular Weight (g/mol) 267.47
MDL Number MFCD00036304

Specifications

Melting Point 217°C
Physical Form Powder
Quantity 100 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula [Co(NH3)6]Cl3
Merck Index 15, 4710
Solubility Information Solubility in water: soluble
Formula Weight 267.47
Percent Purity 99%
Packaging Glass bottle
Chemical Name or Material Hexaamminecobalt(III) chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
In case of inadequate ventilation wear respiratory protection.
IF INHALED: If breathing is di
EINECSNumber 234-103-9

RUO – Research Use Only