accessibility menu, dialog, popup

UserName

H3COONHCH3

CAS RN 105-40-8

IUPAC Name: ethyl N-methylcarbamate
Synonyms: ethyl N-methylcarbamate
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: SURZCVYFPAXNGN-UHFFFAOYSA-N
SMILES: CCOC(=O)NC
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)