missing translation for 'onlineSavingsMsg'
Learn More

Ethyl N-Methylcarbamate 98.0+%, TCI America™

Catalog No. M045625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M045625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M045625G Supplier TCI America Supplier No. M045625G

Chemical Identifiers

CAS 105-40-8
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00041924
InChI Key SURZCVYFPAXNGN-UHFFFAOYSA-N
Synonym ethyl methylcarbamate, n-methylurethane, n-methylurethan, ethyl-n-methylcarbamate, methylcarbamic acid, ethyl ester, n-methyl urethan, carbamic acid, methyl-, ethyl ester, methylurethane van, unii-1w34gcf5cs, ccris 2510
PubChem CID 7752
IUPAC Name ethyl N-methylcarbamate
SMILES CCOC(=O)NC

Specifications

Boiling Point 170°C
Quantity 25 g
Formula Weight 103.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl N-Methylcarbamate
RTECSNumber FC2625000
TSCA Yes