missing translation for 'onlineSavingsMsg'
Learn More

Ethyl methylcarbamate, 99%

Catalog No. AC410260050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC410260050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC410260050 Supplier Thermo Scientific Chemicals Supplier No. 410260050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 105-40-8 | C4H9NO2 | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105-40-8
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00041924
InChI Key SURZCVYFPAXNGN-UHFFFAOYSA-N
Synonym ethyl methylcarbamate, n-methylurethane, n-methylurethan, ethyl-n-methylcarbamate, methylcarbamic acid, ethyl ester, n-methyl urethan, carbamic acid, methyl-, ethyl ester, methylurethane van, unii-1w34gcf5cs, ccris 2510
PubChem CID 7752
IUPAC Name ethyl N-methylcarbamate
SMILES CCOC(=O)NC

Specifications

Boiling Point 170.0°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4180 to 1.4200
Linear Formula CH3NHCOOC2H5
Solubility Information Solubility in water: 6900g/L (16°C). Other solubilities: soluble in alcohol
Density 1.0100g/mL
Quantity 5 g
Flash Point 61°C
Formula Weight 103.12
Percent Purity 99%
Specific Gravity 1.01
Chemical Name or Material Ethyl methylcarbamate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
EINECSNumber 203-295-6

RUO – Research Use Only