accessibility menu, dialog, popup

UserName

CAS RN 10496-18-1

H3CSSCH3

CAS RN 10496-18-1

IUPAC Name: 1-(decyldisulfanyl)decane
Synonyms: 1-(decyldisulfanyl)decane
Molecular Weight (g/mol): 346.68
Molecular Formula: C20H42S2
InChi Key: IDJPKRIELSFBPE-UHFFFAOYSA-N
SMILES: CCCCCCCCCCSSCCCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results