accessibility menu, dialog, popup

UserName

CAS RN 10484-13-6

H3CONOCH3O(E/Z)

CAS RN 10484-13-6

IUPAC Name: 4-{[(4-methoxyphenyl)methylidene]amino}phenyl acetate
Synonyms: 4-{[(4-methoxyphenyl)methylidene]amino}phenyl acetate
Molecular Weight (g/mol): 269.3
Molecular Formula: C16H15NO3
InChi Key: HOYWVKUPOCFKOT-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=NC2=CC=C(OC(C)=O)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results