missing translation for 'onlineSavingsMsg'
Learn More

Periodic Acid, Reagent, 99%, Spectrum™ Chemical

Catalog No. 18605035
Change view
Click to view available options
Quantity:
100 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605035 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18605035 Supplier Spectrum Chemical Mfg Cor Supplier No. P1035100GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

P1035, 10450-60-9, H5IO6

Spectrum™ Chemical Periodic Acid, Reagent

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
MDL Number MFCD00011344
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
IUPAC Name tetrahydroxyiodous acid
SMILES O[I](O)(O)(O)(O)=O

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 99%
Grade Reagent