missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoacrylic acid, 95%

Catalog No. AAL0119414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0119414 25 g
AAL0119406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0119414 Supplier Thermo Scientific Chemicals Supplier No. L0119414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 10443-65-9 | C3H3BrO2 | 150.96 g/mol

2-Bromoacrylic acid is used as a raw material on QSAR models to predict mutagenicity of acrylates. It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoacrylic acid is used as a raw material on QSAR models to predict mutagenicity of acrylates. It is used as a pharmaceutical intermediate.

Solubility
It is miscible with water.

Notes
Heat sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 10443-65-9
Molecular Formula C3H3BrO2
Molecular Weight (g/mol) 150.96
MDL Number MFCD00014333
InChI Key HMENQNSSJFLQOP-UHFFFAOYSA-N
Synonym 2-bromoacrylic acid, bromoacrylic acid, ccris 5478, .alpha.-bromoacrylic acid, 2-bromo-acrylic acid, alpha-bromo acrylic acid, 2-propenoic acid, bromo, acmc-1c0k4, 2-propenoic acid, 2-bromo
PubChem CID 82633
IUPAC Name 2-bromoprop-2-enoic acid
SMILES OC(=O)C(Br)=C

Specifications

Melting Point 65°C to 70°C
Quantity 25 g
UN Number UN3261
Beilstein 1740582
Solubility Information It is miscible with water.
Formula Weight 150.96
Percent Purity 95%
Chemical Name or Material 2-Bromoacrylic acid
Hazard Category H314-H317-H335
Hazard Statement GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 233-928-1
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only