accessibility menu, dialog, popup

UserName

CAS RN 10424-65-4

IUPAC Name: tetramethylazanium pentahydrate hydroxide
Synonyms: tetramethylammonium hydroxide pentahydrate
Molecular Weight (g/mol): 181.23
Molecular Formula: C4H23NO6
InChi Key: MYXKPFMQWULLOH-UHFFFAOYSA-M
SMILES: O.O.O.O.O.[OH-].C[N+](C)(C)C
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (3)
  • (5)
Physical Form 
  • (4)
  • (3)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (9)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (5)
  • (4)
  • (3)
  • (1)
  • (1)