missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium hydroxide pentahydrate, 98%

Catalog No. AAL0965822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0965822 100 g
AAL0965814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0965822 Supplier Thermo Scientific Chemicals Supplier No. L0965822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10424-65-4 | C4H23NO6 | 181.23 g/mol

Tetramethylammonium hydroxide pentahydrate is used in analytical chemistry as an ion pairing agent and a polarographic reagent. It finds application in gas chromatography/mass spectrometry (GC/MS), capillary isotachophoresis and capillary zone electrophoresis and liquid chromatography. Furthermore, it is utilized in the preparation of iron oxide nanoparticles which are useful as magnetic resonance imaging contrast agents and tetramethylammonium dithiocarboxylates. It acts as a developer or an etchant in semiconductor and photoelectric industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethylammonium hydroxide pentahydrate is used in analytical chemistry as an ion pairing agent and a polarographic reagent. It finds application in gas chromatography/mass spectrometry (GC/MS), capillary isotachophoresis and capillary zone electrophoresis and liquid chromatography. Furthermore, it is utilized in the preparation of iron oxide nanoparticles which are useful as magnetic resonance imaging contrast agents and tetramethylammonium dithiocarboxylates. It acts as a developer or an etchant in semiconductor and photoelectric industries.

Solubility
Soluble in water and methanol.

Notes
Air sensitive. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 10424-65-4
Molecular Formula C4H23NO6
Molecular Weight (g/mol) 181.23
MDL Number MFCD00149566
InChI Key MYXKPFMQWULLOH-UHFFFAOYSA-M
Synonym tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm
PubChem CID 82620
IUPAC Name tetramethylazanium;hydroxide;pentahydrate
SMILES O.O.O.O.O.[OH-].C[N+](C)(C)C

Specifications

Melting Point 66°C to 67°C
Linear Formula (CH3)4NOH·5H2O
Quantity 100 g
UN Number UN3423
Beilstein 3714235
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,9224
Solubility Information Soluble in water and methanol.
Formula Weight 181.23 (91.15 Anhydrous)
Percent Purity 98%
Chemical Name or Material Tetramethylammonium hydroxide pentahydrate
Show More Show Less
Hazard Category H300+H310-H314-H335-H370-H372
Hazard Statement GHS H Statement
H300-H310-H370-H372-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Causes damage to organs.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: TETRAMETHYLAMMONIUM HYDROXIDE, SOLID
EINECSNumber 200-882-9
RTECSNumber PA0875000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only