missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Hydroxide Pentahydrate, 98%, Reagents

Catalog No. 01336932
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336932 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336932 Supplier Labchem Inc Supplier No. C23375401KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tetramethylammonium Hydroxide Pentahydrate, 98%, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10424-65-4
Molecular Formula C4H23NO6
Molecular Weight (g/mol) 181.23
MDL Number MFCD00149566
InChI Key MYXKPFMQWULLOH-UHFFFAOYSA-M
IUPAC Name tetramethylazanium pentahydrate hydroxide
SMILES O.O.O.O.O.[OH-].C[N+](C)(C)C

Specifications

Color Colorless/White
Packaging Poly Bottle
Quantity 1 kg
Specific Gravity 1.829
Grade Reagent
Physical Form Solid
Chemical Name or Material Tetramethylammonium Hydroxide Pentahydrate, 98%