missing translation for 'onlineSavingsMsg'
Learn More

all cis-5,8,11,14,17-Eicosapentaenoic Acid 95.0+%, TCI America™

Catalog No. E0441500MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0441500MG 500 mg
E0441100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0441500MG Supplier TCI America Supplier No. E0441500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10417-94-4
Molecular Formula C20H30O2
Molecular Weight (g/mol) 302.46
MDL Number MFCD00065716
InChI Key JAZBEHYOTPTENJ-JLNKQSITSA-N
Synonym EPA, all cis-5,8,11,14,17-Icosapentaenoic Acid
PubChem CID 446284
ChEBI CHEBI:28364
IUPAC Name (5Z,8Z,11Z,14Z,17Z)-icosa-5,8,11,14,17-pentaenoic acid
SMILES CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCCC(O)=O

Specifications

Color Yellow
Quantity 500 mg
Formula Weight 302.46
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material all cis-5,8,11,14,17-Eicosapentaenoic Acid
EINECSNumber (2)-0609
TSCA No
Recommended Storage Freezer