accessibility menu, dialog, popup

UserName

CAS RN 10401-11-3

IUPAC Name: 3-ethynylphenol
Synonyms: 3-ethynylphenol
Molecular Weight (g/mol): 118.14
Molecular Formula: C8H6O
InChi Key: AODMJIOEGCBUQL-UHFFFAOYSA-N
SMILES: OC1=CC=CC(=C1)C#C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results