accessibility menu, dialog, popup

UserName

CAS RN 104-40-5

IUPAC Name: 4-nonylphenol
Synonyms: 4-nonylphenol N-nonylphenol nonylphenol (isomer mixture) P-nonylphenol
Molecular Weight (g/mol): 220.36
Molecular Formula: C15H24O
InChi Key: IGFHQQFPSIBGKE-UHFFFAOYSA-N
SMILES: CCCCCCCCCC1=CC=C(O)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results