accessibility menu, dialog, popup

UserName

CH3ONHNNHNH2S(E)

CAS RN 104-06-3

IUPAC Name: N-{4-[(E)-[(carbamothioylamino)imino]methyl]phenyl}acetamide
Synonyms: thiacetazone thioacetazone
Molecular Weight (g/mol): 236.29
Molecular Formula: C10H12N4OS
InChi Key: SRVJKTDHMYAMHA-WUXMJOGZSA-N
SMILES: CC(=O)NC1=CC=C(\C=N\NC(N)=S)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results