accessibility menu, dialog, popup

UserName

H3COOOHOHOH

CAS RN 1034-01-1

IUPAC Name: octyl 3,4,5-trihydroxybenzoate
Synonyms: octyl gallate caprylyl gallate
Molecular Weight (g/mol): 282.34
Molecular Formula: C15H22O5
InChi Key: NRPKURNSADTHLJ-UHFFFAOYSA-N
SMILES: CCCCCCCCOC(=O)C1=CC(O)=C(O)C(O)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)