missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl gallate, 98+%

Catalog No. AAL0608122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0608122 100 g
AAL0608114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0608122 Supplier Thermo Scientific Chemicals Supplier No. L0608122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1034-01-1 | C15H22O5 | 282.336 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1034-01-1
Molecular Formula C15H22O5
Molecular Weight (g/mol) 282.336
MDL Number MFCD00002197
InChI Key NRPKURNSADTHLJ-UHFFFAOYSA-N
Synonym octyl gallate, n-octyl gallate, progallin o, stabilizer ga 8, n-octylgallate, gallic acid, octyl ester, gallic acid n-octyl ester, gallic acid octyl ester, benzoic acid, 3,4,5-trihydroxy-, octyl ester, oktylester kyseliny gallove
PubChem CID 61253
ChEBI CHEBI:83631
IUPAC Name octyl 3,4,5-trihydroxybenzoate
SMILES CCCCCCCCOC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 96°C to 102°C
Quantity 100 g
Beilstein 2132305
Formula Weight 282.34
Percent Purity ≥98%
Chemical Name or Material n-Octyl gallate
Hazard Category H302-H317
Hazard Statement GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
EINECSNumber 213-853-0
RTECSNumber LW8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only