missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl Gallate 98.0+%, TCI America™

Catalog No. G020625G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G020625G 25 g
G0206250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G020625G Supplier TCI America Supplier No. G020625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1034-01-1
Molecular Formula C15H22O5
Molecular Weight (g/mol) 282.336
MDL Number MFCD00002197
InChI Key NRPKURNSADTHLJ-UHFFFAOYSA-N
Synonym octyl gallate, n-octyl gallate, progallin o, stabilizer ga 8, n-octylgallate, gallic acid, octyl ester, gallic acid n-octyl ester, gallic acid octyl ester, benzoic acid, 3,4,5-trihydroxy-, octyl ester, oktylester kyseliny gallove
PubChem CID 61253
ChEBI CHEBI:83631
IUPAC Name octyl 3,4,5-trihydroxybenzoate
SMILES CCCCCCCCOC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 100°C
Quantity 25 g
Formula Weight 282.34
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material n-Octyl Gallate
EINECSNumber (3)-3071
RTECSNumber LW8225000
TSCA Yes