accessibility menu, dialog, popup

UserName

CAS RN 1033035-83-4

CH3NNH3C

CAS RN 1033035-83-4

IUPAC Name: N2,N7-bis(3-methylphenyl)-N2,N7-diphenyl-9,9'-spirobi[fluorene]-2,7-diamine
Synonyms: N2,N7-bis(3-methylphenyl)-N2,N7-diphenyl-9,9'-spirobi[fluorene]-2,7-diamine
Molecular Weight (g/mol): 678.88
Molecular Formula: C51H38N2
InChi Key: QZTQQBIGSZWRGI-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1)N(C1=CC=CC=C1)C1=CC2=C(C=C1)C1=C(C=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC(C)=C1)C21C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results