missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Bis[N-(m-tolyl)anilino]-9,9'-spirobi[9H-fluorene] 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 213°C |
| Color | Yellow |
| Quantity | 200 mg |
| Formula Weight | 678.88 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,7-Bis[N-(m-tolyl)anilino]-9,9′-spirobi[9H-fluorene] |
Safety and Handling
| TSCA | No |