accessibility menu, dialog, popup

UserName

CAS RN 103-11-7

IUPAC Name: 2-ethylhexyl prop-2-enoate
Synonyms: ethylhexyl acrylate
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: GOXQRTZXKQZDDN-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)C=C
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (6)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)