missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl acrylate, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019107
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019107 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019107 Supplier Merck Emd Millipore Supplier No. 672625MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2-Ethylhexyl acrylates are classified under the ester group of compounds.

Specifications

CAS 103-11-7
Solution Contains 0.001-0.11% monomethyl ether hydroquinone as stabilizer
Vapor Pressure 0.15 mmHg (20°C)
Vapor Density 6.4 (vs. air)
Density 0.884 g/mL (at 20°C)
Physical Form Neat
Linear Formula CH2=CHCOOCH2CH(C2H5)(CH2)3 CH3
MDL Number MFCD00009495
Boiling Point 215°C to 219°C
Synonym (+/-)-Acrylic acid 2-ethylhexyl ester
Quantity 5 mL
Formula Weight 184.28
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less