accessibility menu, dialog, popup

UserName

Last Chance to Save on Spotlight Special Offers

Act Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 1026-92-2

H2COOOOCH2

CAS RN 1026-92-2

IUPAC Name: 1,4-bis(prop-2-en-1-yl) benzene-1,4-dicarboxylate
Synonyms: 1,4-bis(prop-2-en-1-yl) benzene-1,4-dicarboxylate
Molecular Weight (g/mol): 246.26
Molecular Formula: C14H14O4
InChi Key: ZDNFTNPFYCKVTB-UHFFFAOYSA-N
SMILES: C=CCOC(=O)C1=CC=C(C=C1)C(=O)OCC=C
Boiling Point 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.