missing translation for 'onlineSavingsMsg'
Learn More

Diallyl Terephthalate 98.0+%, TCI America™

Catalog No. T148325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T148325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T148325G Supplier TCI America Supplier No. T148325G
Only null left

Chemical Identifiers

CAS 1026-92-2
Molecular Formula C14H14O4
Molecular Weight (g/mol) 246.262
MDL Number MFCD00080462
InChI Key ZDNFTNPFYCKVTB-UHFFFAOYSA-N
Synonym Terephthalic Acid Diallyl Ester
PubChem CID 66102
IUPAC Name bis(prop-2-enyl) benzene-1,4-dicarboxylate
SMILES C=CCOC(=O)C1=CC=C(C=C1)C(=O)OCC=C

Specifications

Color Yellow
Boiling Point 172°C
Quantity 25 g
Formula Weight 246.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diallyl Terephthalate
EINECSNumber (3)-1325&(3)-3847
TSCA Yes
Recommended Storage Refrigerator