accessibility menu, dialog, popup

UserName

CAS RN 102417-94-7

IUPAC Name: 5-(dimethylamino)-N-(4-isothiocyanatophenyl)naphthalene-1-sulfonamide
Synonyms: 5-(dimethylamino)-N-(4-isothiocyanatophenyl)naphthalene-1-sulfonamide
Molecular Weight (g/mol): 383.48
Molecular Formula: C19H17N3O2S2
InChi Key: MZGXHCHKGRQLHR-UHFFFAOYSA-N
SMILES: CN(C)C1=C2C=CC=C(C2=CC=C1)S(=O)(=O)NC1=CC=C(C=C1)N=C=S
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results