accessibility menu, dialog, popup

UserName

CAS RN 102185-33-1

IUPAC Name: disodium 5-bromo-4-chloro-1H-indol-3-yl phosphate
Synonyms: disodium 5-bromo-4-chloro-1H-indol-3-yl phosphate
Molecular Weight (g/mol): 370.43
Molecular Formula: C8H4BrClNNa2O4P
InChi Key: OAZUOCJOEUNDEK-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]P([O-])(=O)OC1=CNC2=CC=C(Br)C(Cl)=C12
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)