accessibility menu, dialog, popup

UserName

ClClClGd

CAS RN 10138-52-0

IUPAC Name: gadolinium(3+) trichloride
Synonyms: gadolinium(III) chloride
Molecular Weight (g/mol): 263.6
Molecular Formula: Cl3Gd
InChi Key: MEANOSLIBWSCIT-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Gd+3]
Grade 
  • (1)
  • (1)
Concentration 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)