missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) Chloride Solution NMR Reference Standard, 1 mg/mL in D2O (99.9 atom % D), DSS 0.1%, Water 1%, MilliporeSigma™ Supelco™

Catalog No. 111002861
Change view
Click to view available options
Quantity:
10 x 0.435 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002861 10 x 0.435 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002861 Supplier Merck Emd Millipore Supplier No. 61317710X8IN

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

NMR Tube Size: 4 mm 8 in.

Specifications

Chemical Name or Material Gadolinium(III) Chloride
Packaging NMR Tube Size: 4 mm 8 in.
CAS 10138-52-0, 7789-20-0
Concentration 1 mg/mL in D2O (99.9 atom% D)
Linear Formula GdCl3
MDL Number MFCD00011024
Industry Type Organic Analysis
Molecular Weight (g/mol) 263.61
Quantity 10 x 0.435 mL
Formula Weight 263.61
Grade NMR Reference Standard
Show More Show Less