missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, puriss., Solstice

Catalog No. 60045651
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
4 x 5 L
Packaging:
Aluminum Bottle
Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045651 5 L Aluminum Bottle
60045646 1 L Glass Bottle
60045647 2.5 L Glass Bottle
60045649 4 x 2.5 L Glass Bottle
60045650 4 x 5 L Aluminum Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60045651 Supplier Solstice Supplier No. 245875L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss., low boiling point hydrogen treated naphtha, meets analytical specification of DAB, bpmin. 75% 40-60 °C (min. 75%)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 101316-46-5
MDL Number MFCD00081849

Specifications

Density 0.642 to 0.656g/mL at 20°C
Quantity 5 L
Flash Point −40°C (−40°F)
Packaging Aluminum Bottle
UN Number UN1268
Grade puriss.
Chemical Name or Material Petroleum Ether, Meets analytical specification of DAB
Hazard Statement P210-P273-P301 + P310-P331-P391-P403 + P235
Precautionary Statement H225-H304-H336-H411
DOTInformation Class 3 - PG 2 - EHS - VP > 110kPa - Petroleum distillates, n.o.s., HI: S
EINECSNumber 309-852-0