missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, puriss. p.a., high boiling, bp 60-80°C, Solstice

Catalog No. 60048551
Change view
Click to view available options
Quantity:
4 x 2.5 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60048551 4 x 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60048551 Supplier Honeywell Inc Supplier No. 322484X2.5L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101316-46-5
MDL Number MFCD00081849

Specifications

Density 0.660 to 0.680g/mL at 20°C
Quantity 4 x 2.5 L
Flash Point −40°C (−40°F)
Packaging Glass Bottle
UN Number UN1268
Grade puriss. p.a.
Chemical Name or Material Petroleum Ether
Hazard Statement P210-P261-P273-P281-P301 + P310-P331
Precautionary Statement H225-H304-H315-H336-H361-H373-H411
DOTInformation class 3 - PG 2 - EHS - VP > 110kPa - Petroleum distillates, n.o.s., HI: S
EINECSNumber 309-852-0