accessibility menu, dialog, popup

UserName

NH2OCl

CAS RN 101-79-1

IUPAC Name: 4-(4-chlorophenoxy)aniline
Synonyms: 4-(4-chlorophenoxy)aniline
Molecular Weight (g/mol): 219.67
Molecular Formula: C12H10ClNO
InChi Key: YTISFYMPVILQRL-UHFFFAOYSA-N
SMILES: NC1=CC=C(OC2=CC=C(Cl)C=C2)C=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)